siRNA Details
Gene Symbol: Tango6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255568.2 | XP_006255630.1 | 4 | 958 | ||
XM_008772562.1 | XP_008770784.1 | 4 | 958 | |||
XM_226428.8 | XP_226428.6 | 4 | 958 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tango6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255568.2 | XP_006255630.1 | 4 | 958 | ||
XM_008772562.1 | XP_008770784.1 | 4 | 958 | |||
XM_226428.8 | XP_226428.6 | 4 | 958 |