siRNA Details
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107425.1 | NP_001100895.1 | 4 | 1460 | ||
XM_003751872.3 | XP_003751920.1 | 3 | 1121 | |||
XM_006255553.2 | XP_006255615.1 | 2 | 3883 | |||
XM_008772503.1 | XP_008770725.1 | 4 | 1175 | |||
XM_008772504.1 | XP_008770726.1 | 2 | 1002 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_018823, NM_001286260, NM_133957, AF369980, AK134022, and others.
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
