siRNA Details
Gene Symbol: Mon1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255674.2 | XP_006255736.1 | 3 | 558 | ||
XM_006255675.2 | XP_006255737.1 | 3 | 372 | |||
XM_008772515.1 | XP_008770737.1 | 2 | 587 |
