siRNA Details
Gene Symbol: Gan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107434.1 | NP_001100904.1 | 2 | 307 | ||
XM_006255685.2 | XP_006255747.1 | 2 | 1089 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107434.1 | NP_001100904.1 | 2 | 307 | ||
XM_006255685.2 | XP_006255747.1 | 2 | 1089 |