siRNA Details
Gene Symbol: Gse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255748.2 | XP_006255810.1 | 13 | 3274 | ||
XM_008772656.1 | XP_008770878.1 | 12 | 9334 | |||
XM_008772657.1 | XP_008770879.1 | 11 | 9217 | |||
XM_008772658.1 | XP_008770880.1 | 14 | 3244 | |||
XM_008772659.1 | XP_008770881.1 | 13 | 4602 | |||
XM_008772660.1 | XP_008770882.1 | 12 | 3055 | |||
XM_008772661.1 | XP_008770883.1 | 12 | 3649 | |||
XM_008772662.1 | XP_008770884.1 | 12 | 4382 | |||
XM_226533.8 | XP_226533.5 | 13 | 3868 |

More Information
Other Species Targeted

