siRNA Details
Gene Symbol: Gse1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006255748.2 | XP_006255810.1 | 15, 16 | 3755 | ||
XM_008772656.1 | XP_008770878.1 | 14, 15 | 9815 | |||
XM_008772657.1 | XP_008770879.1 | 13, 14 | 9698 | |||
XM_008772658.1 | XP_008770880.1 | 16, 17 | 3725 | |||
XM_008772659.1 | XP_008770881.1 | 15, 16 | 5083 | |||
XM_008772660.1 | XP_008770882.1 | 14, 15 | 3536 | |||
XM_008772661.1 | XP_008770883.1 | 14, 15 | 4130 | |||
XM_008772662.1 | XP_008770884.1 | 14, 15 | 4863 | |||
XM_226533.8 | XP_226533.5 | 15, 16 | 4349 |

More Information
Other Species Targeted

