siRNA Details
Gene Symbol: Vps9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107440.2 | NP_001100910.2 | 14, 15 | 1854 | ||
XM_006255767.1 | XP_006255829.1 | 14, 15 | 1900 | |||
XM_008772613.1 | XP_008770835.1 | 14, 15 | 2369 | |||
GenBank mRNA | BC166767.1 | - | 14, 15 | 1854 | 7933213 | |
FQ221344.1 | - | 14, 15 | 1865 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
