siRNA Details
Gene Symbol: Ccsap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107441.1 | NP_001100911.1 | 2 | 548 | ||
XM_006255811.2 | XP_006255873.1 | 3 | 1048 | |||
GenBank mRNA | FQ211944.1 | - | 2 | 322 | ||
FQ212467.1 | - | 2 | 321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
