siRNA Details
Gene Symbol: Fam204a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107448.2 | NP_001100918.1 | 5 | 558 | ||
XM_006231673.2 | XP_006231735.1 | 5 | 1374 |

More Information
Other Species Targeted

