siRNA Details
Gene Symbol: Fam45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127681.1 | NP_001121153.1 | 2 | 195 | ||
XM_006231682.1 | XP_006231744.1 | 2 | 187 | |||
GenBank mRNA | BC167081.1 | - | 2 | 164 |

More Information
Other Species Targeted

