siRNA Details
Gene Symbol: Fam45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127681.1 | NP_001121153.1 | 2 | 136 | ||
XM_006231682.1 | XP_006231744.1 | 2 | 128 | |||
GenBank mRNA | BC167081.1 | - | 2 | 105 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
