siRNA Details
Gene Symbol: Rnf146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012060.2 | NP_001012060.2 | 3 | 717 | ||
XM_006227744.2 | XP_006227806.1 | 3 | 774 | |||
XM_006227745.2 | XP_006227807.1 | 4 | 822 | |||
GenBank mRNA | BC083675.1 | - | 3 | 717 | 7135728 |
