siRNA Details
Gene Symbol: Cep72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006227800.2 | XP_006227862.1 | 2 | 207 | ||
XM_006227801.2 | XP_006227863.1 | 2 | 207 | |||
XM_008758697.1 | XP_008756919.1 | 2 | 103 | |||
XM_217737.8 | XP_217737.4 | 2 | 209 |
