siRNA Details
Gene Symbol: Nkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107454.1 | NP_001100924.1 | 7 | 711 | ||
XM_008758691.1 | XP_008756913.1 | 3 | 1409 | |||
XM_008758692.1 | XP_008756914.1 | 10 | 2445 | |||
XM_008758693.1 | XP_008756915.1 | 7 | 1226 |
