siRNA Details
Gene Symbol: Acat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006995.1 | NP_001006996.1 | 5, 6 | 667 | ||
GenBank mRNA | AY325187.1 | - | 641 | |||
BC083872.1 | - | 5, 6 | 667 | 7128564 |

More Information
Other Species Targeted

Gene Ontology Categories:
