siRNA Details
Gene Symbol: Cnksr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012061.1 | NP_001012061.1 | 13 | 1933 | ||
XM_006227875.2 | XP_006227937.1 | 13 | 3090 | |||
GenBank mRNA | AY328890.1 | - | 13 | 1933 | ||
AY333962.1 | - | 5 | 566 |
