siRNA Details
Gene Symbol: Cnksr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012061.1 | NP_001012061.1 | 11 | 1762 | ||
XM_006227875.2 | XP_006227937.1 | 11 | 2919 | |||
GenBank mRNA | AY328890.1 | - | 11 | 1762 | ||
AY333962.1 | - | 3 | 392 |
