siRNA Details
Gene Symbol: Zbtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107460.1 | NP_001100930.1 | 1 | 154 | ||
XM_008758705.1 | XP_008756927.1 | 1 | 153 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001033466, AK132308, AK160748, AK030936.
Gene Ontology Categories:
