siRNA Details
Gene Symbol: Dact2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107464.2 | NP_001100934.2 | 4 | 1845 | ||
XM_006227960.2 | XP_006228022.1 | 4 | 1776 | |||
XM_006227962.2 | XP_006228024.1 | 4 | 2295 | |||
XM_006227964.2 | XP_006228026.1 | 4 | 1900 | |||
XM_006227965.2 | XP_006228027.1 | 4 | 2286 | |||
XM_008758760.1 | XP_008756982.1 | 4 | 2324 | |||
GenBank mRNA | CK473696.1 | - | 1 | 532 | ||
FQ215550.1 | - | 1 | 514 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
