siRNA Details
Gene Symbol: Fam120b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107466.3 | NP_001100936.2 | 2 | 744 | ||
XM_006231141.2 | XP_006231203.1 | 2 | 873 | |||
XM_006231142.2 | XP_006231204.1 | 2 | 872 | |||
XM_008758761.1 | XP_008756983.1 | 2 | 585 | |||
GenBank mRNA | FQ210918.1 | - | 2 | 744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
