siRNA Details
Gene Symbol: Zfp53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107468.1 | NP_001100938.1 | 5 | 1563 | ||
XM_008758763.1 | XP_008756985.1 | 5 | 1572 |

More Information
Other Species Targeted

Gene Ontology Categories:
