siRNA Details
Gene Symbol: Zc3h12d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107469.1 | NP_001100939.1 | 6 | 1527 | ||
XM_006227616.2 | XP_006227678.1 | 6 | 2485 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zc3h12d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107469.1 | NP_001100939.1 | 6 | 1527 | ||
XM_006227616.2 | XP_006227678.1 | 6 | 2485 |