siRNA Details
Gene Symbol: Tab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012062.1 | NP_001012062.1 | 4 | 330 | ||
XM_006227617.2 | XP_006227679.1 | 3 | 363 | |||
XM_006227618.2 | XP_006227680.1 | 3 | 709 | |||
XM_006227620.2 | XP_006227682.1 | 3 | 360 | |||
XM_008758608.1 | XP_008756830.1 | 4 | 427 | |||
GenBank mRNA | BC085788.1 | - | 4 | 330 | 7108689 |
