siRNA Details
Gene Symbol: Phactr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_214458.1 | NP_999623.1 | 11 | 1678 | ||
XM_008758610.1 | XP_008756832.1 | 12 | 2835 | |||
XM_008758611.1 | XP_008756833.1 | 12 | 2608 | |||
GenBank mRNA | AY500158.1 | - | 11 | 1678 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
