siRNA Details
Gene Symbol: Gpr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025680.1 | NP_001020851.1 | 1 | 846 | ||
XM_006228418.1 | XP_006228480.1 | 2 | 1437 | |||
XM_006228419.1 | XP_006228481.1 | 2 | 2761 | |||
GenBank mRNA | BC099196.1 | - | 1 | 846 | 7383735 |
