siRNA Details
Gene Symbol: Eif2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017489.1 | NP_001017489.1 | 13 | 1642 | ||
XM_006249790.2 | XP_006249852.1 | 13 | 1666 | |||
GenBank mRNA | BC087701.1 | - | 13 | 1642 | 7314085 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
