siRNA Details
Gene Symbol: Cic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008758933.1 | XP_008757155.1 | 2 | 2272 | ||
XM_008758934.1 | XP_008757156.1 | 2 | 2536 | |||
XM_008758935.1 | XP_008757157.1 | 2 | 2272 | |||
XM_008758936.1 | XP_008757158.1 | 2 | 2272 | |||
XM_008758937.1 | XP_008757159.1 | 2 | 2272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
