siRNA Details
Gene Symbol: B3gnt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107492.1 | NP_001100962.1 | 3 | 1113 | ||
XM_006228433.2 | XP_006228495.1 | 2 | 2511 | |||
XM_006228434.2 | XP_006228496.1 | 2 | 2553 | |||
XM_006228436.2 | XP_006228498.1 | 2 | 2199 |

More Information
Other Species Targeted

Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
