siRNA Details
Gene Symbol: Exosc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107493.2 | NP_001100963.2 | 2 | 279 | ||
XM_006228437.2 | XP_006228499.1 | 3 | 320 | |||
GenBank mRNA | BC167023.1 | - | 2 | 191 | ||
FQ225297.1 | - | 2 | 203 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_138586, BY198385, AK155228, BY196593, AK007372, and others.
Gene Ontology Categories:
