siRNA Details
Gene Symbol: Exosc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107493.2 | NP_001100963.2 | 1, 2 | 232 | ||
XM_006228437.2 | XP_006228499.1 | 2, 3 | 273 | |||
GenBank mRNA | BC167023.1 | - | 1, 2 | 144 | ||
FQ225297.1 | - | 1, 2 | 156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
