siRNA Details
Gene Symbol: Itpkc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178094.3 | NP_835195.1 | 1 | 1021 | ||
GenBank mRNA | AJ440782.1 | - | 1 | 1021 | ||
AY160770.1 | - | 1 | 1267 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
