siRNA Details
Gene Symbol: Supt5h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107497.2 | NP_001100967.2 | 17 | 1487 | ||
GenBank mRNA | BC166741.1 | - | 15 | 2400 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
