siRNA Details
Gene Symbol: Spred3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173344.1 | NP_001166815.1 | 2 | 627 | ||
XM_008759144.1 | XP_008757366.1 | 3 | 265 | |||
XM_008759145.1 | XP_008757367.1 | 3 | 212 | |||
XM_008759146.1 | XP_008757368.1 | 3 | 334 | |||
GenBank mRNA | BC166515.1 | - | 2 | 627 | 7131550 |

More Information
Other Species Targeted

Gene Ontology Categories:
