siRNA Details
Gene Symbol: Wdr62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191623.2 | NP_001178552.2 | 32 | 4380 | ||
XM_006228797.1 | XP_006228859.1 | 31 | 4367 | |||
GenBank mRNA | BC087154.1 | - | 11 | 1819 | ||
BC104681.1 | - | 12 | 1948 |
Gene Symbol: LOC103689973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759279.1 | XP_008757501.1 | 31 | 4446 |
