siRNA Details
Gene Symbol: Cd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107503.1 | NP_001100973.1 | 4 | 801 | ||
XM_006228828.2 | XP_006228890.1 | 6 | 782 | |||
XM_008759152.1 | XP_008757374.1 | 5 | 855 | |||
XM_008759153.1 | XP_008757375.1 | 5 | 616 | |||
XM_008759154.1 | XP_008757376.1 | 5 | 616 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
