siRNA Details
Gene Symbol: Chst8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107504.2 | NP_001100974.1 | 4 | 775 | ||
XM_006228853.2 | XP_006228915.1 | 4 | 1660 | |||
XM_006228854.2 | XP_006228916.1 | 4 | 1086 | |||
GenBank mRNA | BC161905.1 | - | 4 | 775 | 8367390 |
