siRNA Details
Gene Symbol: Dpy19l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135835.2 | NP_001129307.2 | 4 | 507 | ||
XM_006228888.2 | XP_006228950.1 | 4 | 999 | |||
XM_006228889.2 | XP_006228951.1 | 4 | 409 | |||
XM_006228890.2 | XP_006228952.1 | 4 | 1086 | |||
XM_008759162.1 | XP_008757384.1 | 2 | 113 | |||
XM_008759163.1 | XP_008757385.1 | 2 | 59 |
