siRNA Details
Gene Symbol: Tshz3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107506.1 | NP_001100976.1 | 2 | 3038 | ||
XM_006228893.1 | XP_006228955.1 | 2 | 3074 |

More Information
Other Species Targeted

Gene Ontology Categories:
