siRNA Details
Gene Symbol: Acpt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107510.1 | NP_001100980.1 | 5 | 514 | ||
XM_006229051.2 | XP_006229113.1 | 7 | 862 | |||
XM_006229052.2 | XP_006229114.1 | 8 | 783 | |||
XM_006229053.2 | XP_006229115.1 | 7 | 898 | |||
XM_006229054.2 | XP_006229116.1 | 7 | 934 | |||
XM_008759366.1 | XP_008757588.1 | 6 | 675 | |||
XM_008759367.1 | XP_008757589.1 | 7 | 898 | |||
XM_008759368.1 | XP_008757590.1 | 7 | 895 | |||
XM_008759369.1 | XP_008757591.1 | 6 | 1727 | |||
GenBank mRNA | BC168932.1 | - | 7 | 939 |

More Information
Other Species Targeted

Gene Ontology Categories:
