siRNA Details
Gene Symbol: Cpt1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034925.2 | NP_001030097.2 | 17 | 2294 | ||
XM_006229066.2 | XP_006229128.1 | 17 | 3316 | |||
XM_008759376.1 | XP_008757598.1 | 16 | 3460 | |||
XM_008759377.1 | XP_008757599.1 | 15 | 2297 | |||
GenBank mRNA | BC105882.1 | - | 8 | 1057 |
