siRNA Details
Gene Symbol: Tmem143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107513.1 | NP_001100983.1 | 5 | 681 | ||
XM_006229089.2 | XP_006229151.1 | 5 | 676 | |||
XM_006229090.2 | XP_006229152.1 | 4 | 542 | |||
XM_006229092.2 | XP_006229154.1 | 5 | 647 | |||
XM_008759386.1 | XP_008757608.1 | 5 | 647 |

More Information
Other Species Targeted

Gene Ontology Categories:
