siRNA Details
Gene Symbol: Tmem143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107513.1 | NP_001100983.1 | 4, 5 | 659 | ||
XM_006229089.2 | XP_006229151.1 | 4, 5 | 654 | |||
XM_006229090.2 | XP_006229152.1 | 3, 4 | 520 | |||
XM_006229092.2 | XP_006229154.1 | 4, 5 | 625 | |||
XM_008759386.1 | XP_008757608.1 | 4, 5 | 625 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
