siRNA Details
Gene Symbol: Smox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235063.2 | XP_006235125.1 | 7 | 1787 | ||
XM_008762165.1 | XP_008760387.1 | 7 | 1803 |

More Information
Other Species Targeted

Gene Ontology Categories:
