siRNA Details
Gene Symbol: Smox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134854.1 | NP_001128326.1 | 6 | 2233 | ||
XM_006235063.2 | XP_006235125.1 | 8 | 2215 | |||
XM_006235064.2 | XP_006235126.1 | 7 | 2124 | |||
XM_008762165.1 | XP_008760387.1 | 8 | 2231 | |||
GenBank mRNA | BC161894.1 | - | 6 | 2233 | 7193521 |
