siRNA Details
Gene Symbol: Nipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107519.1 | NP_001100989.1 | 1, 2 | 175 | ||
XM_008759394.1 | XP_008757616.1 | 1, 2 | 206 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107519.1 | NP_001100989.1 | 1, 2 | 175 | ||
XM_008759394.1 | XP_008757616.1 | 1, 2 | 206 |