siRNA Details
Gene Symbol: Herc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107520.1 | NP_001100990.1 | 13 | 1819 | ||
XM_006229300.2 | XP_006229362.1 | 14 | 2001 | |||
XM_006229301.2 | XP_006229363.1 | 14 | 2029 | |||
XM_006229302.2 | XP_006229364.1 | 14 | 1986 | |||
XM_006229304.2 | XP_006229366.1 | 14 | 2001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
