siRNA Details
Gene Symbol: Ttc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025681.1 | NP_001020852.2 | 7 | 921 | ||
XM_008759515.1 | XP_008757737.1 | 7 | 1813 | |||
XM_008759516.1 | XP_008757738.1 | 7 | 1340 | |||
XM_008759518.1 | XP_008757740.1 | 6 | 1632 | |||
GenBank mRNA | BC097953.1 | - | 7 | 921 | 7455883 |
