siRNA Details
Gene Symbol: Man2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107527.1 | NP_001100997.1 | 21 | 3201 | ||
XM_008759527.1 | XP_008757749.1 | 21 | 3302 | |||
XM_008759528.1 | XP_008757750.1 | 22 | 3359 | |||
XM_008759529.1 | XP_008757751.1 | 20 | 3114 | |||
XM_008759530.1 | XP_008757752.1 | 21 | 3300 |

More Information
Other Species Targeted

Gene Ontology Categories:
