siRNA Details
Gene Symbol: Hddc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107528.1 | NP_001100998.1 | 2 | 146 | ||
XM_006229431.1 | XP_006229493.1 | 2 | 272 | |||
XM_008759531.1 | XP_008757753.1 | 2 | 169 |

More Information
Other Species Targeted

