siRNA Details
Gene Symbol: Pde8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198767.1 | NP_942062.1 | 12, 13 | 1158 | ||
XM_006229438.2 | XP_006229500.1 | 12, 13 | 1345 | |||
XM_006229439.2 | XP_006229501.1 | 9, 10 | 1156 | |||
GenBank mRNA | AB092696.1 | - | 12, 13 | 1158 |
